Product Details

SNP ID
rs182823850
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58422705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTAGTGTGTTTATCAGAGCAA[G/T]ACAGAGCCATGACTGCTGGAACATC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR5B2 PubMed Links

Gene Details

Gene
OR5B2
Gene Name
olfactory receptor family 5 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005566.2 609 Missense Mutation NP_001005566.1

View Full Product Details