Product Details
- SNP ID
-
rs182482982
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52515039 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGCCAAAGCCCACCCCCAGCCCT[C/T]GGCCACTGCTTGCACTGAAGCCAGA
- Phenotype
-
MIM: 148040
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT5
PubMed Links
Gene Details
- Gene
- KRT5
- Gene Name
- keratin 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000424.3 |
1839 |
Missense Mutation |
|
|
NP_000415.2 |
View Full Product Details