Product Details

SNP ID
rs182578557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23097878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCAGAAAAATTACAACCACTG[C/T]TGGATAGTCTGGAGCAGCTTAGTGT
Phenotype
MIM: 604562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACIN1 PubMed Links

Gene Details

Gene
ACIN1
Gene Name
apoptotic chromatin condensation inducer 1
There are no transcripts associated with this gene.

Gene
C14orf119
Gene Name
chromosome 14 open reading frame 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017924.3 367 Silent Mutation CTG,TTG L74L NP_060394.1
XM_017021390.1 367 Silent Mutation CTG,TTG L74L XP_016876879.1
Gene
LOC100128908
Gene Name
leishmanolysin homolog
There are no transcripts associated with this gene.

View Full Product Details