Product Details

SNP ID
rs182595261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1634976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGGCCCCAGGAAGCCCAGCTT[C/T]TGGGGCTGAGTTGGGGCTCTGGCCG
Phenotype
MIM: 607873 MIM: 610791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARF1 PubMed Links

Gene Details

Gene
SCARF1
Gene Name
scavenger receptor class F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003693.3 1268 Missense Mutation AAA,GAA K759E NP_003684.2
NM_145350.2 1268 UTR 3 NP_663325.1
XM_017025256.1 1268 Missense Mutation AAA,GAA K348E XP_016880745.1
Gene
SLC43A2
Gene Name
solute carrier family 43 member 2
There are no transcripts associated with this gene.

View Full Product Details