Product Details

SNP ID
rs183283680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137871290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATATGCGGAAAGCTGTGAAGAT[A/T]CGGGAGAGAACTCCAGAAGACATTT
Phenotype
MIM: 191163
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130476 PubMed Links

Gene Details

Gene
LOC100130476
Gene Name
uncharacterized LOC100130476
There are no transcripts associated with this gene.

Gene
LOC107986649
Gene Name
uncharacterized LOC107986649
There are no transcripts associated with this gene.

Gene
TNFAIP3
Gene Name
TNF alpha induced protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270507.1 409 Silent Mutation ATA,ATT I21I NP_001257436.1
NM_001270508.1 409 Silent Mutation ATA,ATT I21I NP_001257437.1
NM_006290.3 409 Silent Mutation ATA,ATT I21I NP_006281.1
XM_005267119.1 409 Silent Mutation ATA,ATT I21I XP_005267176.1
XM_006715555.1 409 Intron XP_006715618.1
XM_011536095.1 409 Silent Mutation ATA,ATT I21I XP_011534397.1
XM_011536096.1 409 Silent Mutation ATA,ATT I21I XP_011534398.1

View Full Product Details