Product Details

SNP ID
rs183234371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99885566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTAGGTGTTATAGTAACAGAT[A/G]ATACTTTTATTTTTGGAAAGTCCAA
Phenotype
MIM: 603296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR3 PubMed Links

Gene Details

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 462 Silent Mutation ATC,ATT I64I NP_001230665.1
NM_021970.3 462 Silent Mutation ATC,ATT I71I NP_068805.1

View Full Product Details