Product Details

SNP ID
rs183365581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67149775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGCCAGGAACTCCAGGAGATT[G/T]CCCACGTTCACGAACACATCTGCGT
Phenotype
MIM: 603500
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT9 PubMed Links

Gene Details

Gene
B3GNT9
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033309.2 1032 Silent Mutation GGA,GGC G237G NP_171608.2
Gene
C16orf70
Gene Name
chromosome 16 open reading frame 70
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
There are no transcripts associated with this gene.

View Full Product Details