Product Details

SNP ID
rs183963488
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:35816969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGACTGTTAGCCAATCATTTTC[A/C]GGTTCAGATTCCTAAAATAGATGTG
Phenotype
MIM: 607356
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGO4 PubMed Links

Gene Details

Gene
AGO4
Gene Name
argonaute 4, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017629.3 182 Missense Mutation CAG,CCG Q36P NP_060099.2
XM_005270578.3 182 UTR 5 XP_005270635.1
XM_005270579.3 182 Intron XP_005270636.1
XM_011540883.2 182 Intron XP_011539185.1
XM_011540884.2 182 Intron XP_011539186.1
XM_017000529.1 182 Missense Mutation CAG,CCG Q36P XP_016856018.1

View Full Product Details