Product Details

SNP ID
rs183472896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6621945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTGGAGCCCGAGGAGGAGGA[A/C]GAGGTGGAGCTGGAGCTCATGCCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF853 PubMed Links

Gene Details

Gene
ZNF853
Gene Name
zinc finger protein 853
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017560.1 1395 Missense Mutation GAA,GAC E318D NP_060030.1
XM_011515438.2 1395 Missense Mutation GAA,GAC E344D XP_011513740.1
XM_011515439.2 1395 Missense Mutation GAA,GAC E329D XP_011513741.1
XM_017012371.1 1395 Missense Mutation GAA,GAC E303D XP_016867860.1

View Full Product Details