Product Details

SNP ID
rs184105229
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:224190042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCATCAACCCCAAACCAATTA[C/T]GTATCTGGAAGTTATCAATACCGTG
Phenotype
MIM: 615843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEGS1 PubMed Links

Gene Details

Gene
DEGS1
Gene Name
delta 4-desaturase, sphingolipid 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321541.1 677 Missense Mutation ACG,ATG T183M NP_001308470.1
NM_001321542.1 677 Missense Mutation ACG,ATG T147M NP_001308471.1
NM_003676.3 677 Missense Mutation ACG,ATG T183M NP_003667.1
XM_017002648.1 677 Missense Mutation ACG,ATG T147M XP_016858137.1

View Full Product Details