Product Details

SNP ID
rs184228156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10802132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCGTTCAAATTCTCAGTGGCTG[C/G]AAGGCTAATAAACACAGAGAGAACC
Phenotype
MIM: 604793 MIM: 604794 MIM: 604795
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R7 PubMed Links

Gene Details

Gene
TAS2R7
Gene Name
taste 2 receptor member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023919.2 496 Missense Mutation CCA,GCA P147A NP_076408.1
Gene
TAS2R8
Gene Name
taste 2 receptor member 8
There are no transcripts associated with this gene.

Gene
TAS2R9
Gene Name
taste 2 receptor member 9
There are no transcripts associated with this gene.

View Full Product Details