Product Details
- SNP ID
-
rs184266726
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:113895339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGGTGTCAGGATGGAATTCTCCC[C/T]GCCAAGGCAACACTTGCTGATGAGC
- Phenotype
-
MIM: 607245
MIM: 609683
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AP4B1
PubMed Links
Gene Details
- Gene
- AP4B1
- Gene Name
- adaptor related protein complex 4 beta 1 subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001253852.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q649R |
NP_001240781.1 |
NM_001253853.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q550R |
NP_001240782.1 |
NM_001308312.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q481R |
NP_001295241.1 |
NM_006594.4 |
1648 |
Missense Mutation |
CAG,CGG |
Q649R |
NP_006585.2 |
XM_011540523.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q574R |
XP_011538825.1 |
XM_011540524.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q574R |
XP_011538826.1 |
XM_011540525.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q556R |
XP_011538827.1 |
XM_011540528.2 |
1648 |
Missense Mutation |
CAG,CGG |
Q324R |
XP_011538830.1 |
XM_017000088.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q610R |
XP_016855577.1 |
XM_017000089.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q545R |
XP_016855578.1 |
XM_017000090.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q481R |
XP_016855579.1 |
XM_017000091.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q452R |
XP_016855580.1 |
XM_017000092.1 |
1648 |
Missense Mutation |
CAG,CGG |
Q324R |
XP_016855581.1 |
XM_017000093.1 |
1648 |
Intron |
|
|
XP_016855582.1 |
- Gene
- AP4B1-AS1
- Gene Name
- AP4B1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- BCL2L15
- Gene Name
- BCL2 like 15
There are no transcripts associated with this gene.
- Gene
- DCLRE1B
- Gene Name
- DNA cross-link repair 1B
There are no transcripts associated with this gene.
View Full Product Details