Product Details

SNP ID
rs184740259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56252967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTTCATTCTGTTGTCCATTCT[A/G]AAGATGCATTCTGCTAAGGGAAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5T3 PubMed Links

Gene Details

Gene
OR5T3
Gene Name
olfactory receptor family 5 subfamily T member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004747.1 768 Silent Mutation CTA,CTG L256L NP_001004747.1

View Full Product Details