Product Details

SNP ID
rs185464452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159800791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGTCTGGACATAGAGTTCAC[A/G]GGCCTTCGTTCTAACCTGTCTGGGC
Phenotype
MIM: 611831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL18 PubMed Links

Gene Details

Gene
MRPL18
Gene Name
mitochondrial ribosomal protein L18
There are no transcripts associated with this gene.

Gene
PNLDC1
Gene Name
PARN like, ribonuclease domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271862.1 181 Silent Mutation ACA,ACG T32T NP_001258791.1
NM_173516.2 181 Silent Mutation ACA,ACG T21T NP_775787.1
XM_011535491.2 181 Silent Mutation ACA,ACG T21T XP_011533793.1
XM_011535493.2 181 Intron XP_011533795.1
XM_017010312.1 181 Silent Mutation ACA,ACG T64T XP_016865801.1
XM_017010313.1 181 Silent Mutation ACA,ACG T64T XP_016865802.1
XM_017010314.1 181 Silent Mutation ACA,ACG T64T XP_016865803.1
XM_017010315.1 181 Silent Mutation ACA,ACG T32T XP_016865804.1
XM_017010316.1 181 Silent Mutation ACA,ACG T21T XP_016865805.1

View Full Product Details