Product Details

SNP ID
rs185431381
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:241660545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATGCAGATGCAATTGGTTATG[A/G]CAAGTTTTATGCATCGGTACAGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR64 PubMed Links

Gene Details

Gene
WDR64
Gene Name
WD repeat domain 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144625.4 368 Missense Mutation GAC,GGC D54G NP_653226.4
XM_006711736.3 368 Missense Mutation GAC,GGC D54G XP_006711799.1
XM_011544085.2 368 Intron XP_011542387.1
XM_011544086.2 368 Intron XP_011542388.1
XM_011544087.2 368 Missense Mutation GAC,GGC D54G XP_011542389.1
XM_011544091.1 368 Missense Mutation GAC,GGC D54G XP_011542393.1
XM_011544092.2 368 Missense Mutation GAC,GGC D54G XP_011542394.1
XM_017000315.1 368 Intron XP_016855804.1

View Full Product Details