Product Details

SNP ID
rs185556019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3053864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCCTGGTGGGTGTCACCATC[A/G]TGCCCGTTCTTGTCTTCCTTGGAGA
Phenotype
MIM: 600188 MIM: 601041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AES PubMed Links

Gene Details

Gene
AES
Gene Name
amino-terminal enhancer of split
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130.5 719 Silent Mutation NP_001121.2
NM_198969.1 719 Silent Mutation NP_945320.1
NM_198970.1 719 Silent Mutation NP_945321.1
XM_006722664.1 719 Silent Mutation XP_006722727.1
Gene
TLE2
Gene Name
transducin like enhancer of split 2
There are no transcripts associated with this gene.

View Full Product Details