Product Details

SNP ID
rs185222252
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:2185400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTTCCGATGATGTGGAAGCTC[C/T]TCTGATGCCTTTTTTCCGTGAGTGG
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 873 Missense Mutation AGA,GGA R191G NP_001139782.1
NM_001256945.1 873 Missense Mutation AGA,GGA R150G NP_001243874.1
NM_001256946.1 873 Intron NP_001243875.1
NM_001256947.1 873 Intron NP_001243876.1
NM_001282670.1 873 Intron NP_001269599.1
NM_001282671.1 873 Missense Mutation AGA,GGA R108G NP_001269600.1
NM_001282672.1 873 Missense Mutation AGA,GGA R108G NP_001269601.1
NM_001282673.1 873 UTR 3 NP_001269602.1
NM_182533.2 873 Intron NP_872339.2
XM_006710419.3 873 Intron XP_006710482.1
XM_006710421.3 873 Intron XP_006710484.1
XM_011540914.2 873 Intron XP_011539216.1
XM_011540921.2 873 Intron XP_011539223.1
XM_011540922.1 873 Intron XP_011539224.1
XM_017000553.1 873 Intron XP_016856042.1
XM_017000554.1 873 UTR 3 XP_016856043.1
XM_017000555.1 873 Missense Mutation AGA,GGA R108G XP_016856044.1
XM_017000556.1 873 Missense Mutation AGA,GGA R108G XP_016856045.1
XM_017000557.1 873 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 873 Intron NP_001028753.1
NM_001033582.1 873 Intron NP_001028754.1
NM_001242874.1 873 Intron NP_001229803.1
NM_002744.4 873 Intron NP_002735.3
XM_011541773.1 873 Intron XP_011540075.1
XM_011541774.1 873 Intron XP_011540076.1
XM_011541775.2 873 Intron XP_011540077.1
XM_011541776.1 873 Intron XP_011540078.1
XM_011541778.1 873 Intron XP_011540080.1
XM_017001789.1 873 Intron XP_016857278.1
XM_017001790.1 873 Intron XP_016857279.1
XM_017001791.1 873 Intron XP_016857280.1
XM_017001792.1 873 Intron XP_016857281.1
XM_017001793.1 873 Intron XP_016857282.1
XM_017001794.1 873 Intron XP_016857283.1
XM_017001795.1 873 Intron XP_016857284.1
XM_017001796.1 873 Intron XP_016857285.1
XM_017001797.1 873 Intron XP_016857286.1
XM_017001798.1 873 Intron XP_016857287.1
XM_017001799.1 873 Intron XP_016857288.1
XM_017001800.1 873 Intron XP_016857289.1
XM_017001801.1 873 Intron XP_016857290.1
XM_017001802.1 873 Intron XP_016857291.1
XM_017001803.1 873 Intron XP_016857292.1
XM_017001804.1 873 Intron XP_016857293.1
XM_017001805.1 873 Intron XP_016857294.1

View Full Product Details