Product Details

SNP ID
rs185412063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAGAAAATCAAGAGCAAAACA[G/T]CAAAAAGGAAGATAAGCAATGGCAC
Phenotype
MIM: 604796
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 897 Missense Mutation NP_062545.1

View Full Product Details