Product Details

SNP ID
rs186392388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37089060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTCTTTATTATTGCCAACATC[C/T]TAGAAACATAGTAAGTCTAAACTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPATCH11 PubMed Links

Gene Details

Gene
GPATCH11
Gene Name
G-patch domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278505.1 Intron NP_001265434.1
NM_001322249.1 Intron NP_001309178.1
NM_174931.3 Intron NP_777591.3
XM_017003748.1 Intron XP_016859237.1
Gene
HEATR5B
Gene Name
HEAT repeat containing 5B
There are no transcripts associated with this gene.

View Full Product Details