Product Details

SNP ID
rs186220740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29086826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGAGAAGGCTGCCATCCTTAG[A/G]CAGAACCAATAATTCATGATGACTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2B3 PubMed Links

Gene Details

Gene
OR2B3
Gene Name
olfactory receptor family 2 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005226.2 488 Silent Mutation TGC,TGT C141C NP_001005226.1

View Full Product Details