Product Details

SNP ID
rs187130387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24428878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACCTGCATTGTAGGTGATGCTG[C/T]CGAATGAATACCCAGGTACAGCTCC
Phenotype
MIM: 615319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPACT PubMed Links

Gene Details

Gene
IMPACT
Gene Name
impact RWD domain protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018439.3 287 Missense Mutation CCG,TCG P59S NP_060909.1
XM_017025832.1 287 Missense Mutation CCG,TCG P41S XP_016881321.1

View Full Product Details