Product Details

SNP ID
rs187134302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111875174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTTCTTCTGATCTATCCAAAA[A/G]CTAACATCATGCACATAACATGGAG
Phenotype
MIM: 606941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG9 PubMed Links

Gene Details

Gene
ALG9
Gene Name
ALG9, alpha-1,2-mannosyltransferase
There are no transcripts associated with this gene.

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
There are no transcripts associated with this gene.

Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138378.2 1948 Silent Mutation AGC,AGT S541S NP_612387.1

View Full Product Details