Product Details
- SNP ID
-
rs186726581
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:132404757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCAGCTGGGGTCCAGCTCCCCCC[C/G]TCCTGCCCGCGGGGTTCAGCCCCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PWWP2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10747057] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PWWP2B
- Gene Name
- PWWP domain containing 2B
View Full Product Details