Product Details

SNP ID
rs187771697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51058590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCGGCACACAACATGTTGTCA[C/G]TGATCTTTCCTGGGTAGACTTGACG
Phenotype
MIM: 605505
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLK13 PubMed Links

Gene Details

Gene
KLK13
Gene Name
kallikrein related peptidase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015596.1 636 Missense Mutation ACT,AGT T198S NP_056411.1

View Full Product Details