Product Details

SNP ID
rs187788865
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169840103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTCCTATCTCAGTGATCTGA[A/G]GAAGCTTGGTCACAGCATATAACAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LRRC31 PubMed Links

Gene Details

Gene
LRRC31
Gene Name
leucine rich repeat containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277127.1 1387 UTR 3 NP_001264056.1
NM_001277128.1 1387 Missense Mutation CCT,CTT P457L NP_001264057.1
NM_024727.3 1387 Missense Mutation CCT,CTT P513L NP_079003.2
XM_011513158.2 1387 Missense Mutation CCT,CTT P457L XP_011511460.1
XM_011513159.2 1387 Missense Mutation CCT,CTT P457L XP_011511461.1
XM_011513160.2 1387 Intron XP_011511462.1
XM_017007204.1 1387 Missense Mutation CCT,CTT P448L XP_016862693.1
Gene
LRRIQ4
Gene Name
leucine rich repeats and IQ motif containing 4
There are no transcripts associated with this gene.

View Full Product Details