Product Details
- SNP ID
-
rs187882824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:28004054 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTTAAAACTAAATTGTTCCTCT[A/G]CGAATCTTCGACCTGAGCCTGCGGA
- Phenotype
-
MIM: 136351
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLT3
PubMed Links
Gene Details
- Gene
- FLT3
- Gene Name
- fms related tyrosine kinase 3
There are no transcripts associated with this gene.
View Full Product Details