Product Details

SNP ID
rs187898853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39342991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAATATAGAAATAAATGTGGTA[C/T]AGCACAGTACAATAATGGACAAAAG
Phenotype
MIM: 606710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHFP PubMed Links

Gene Details

Gene
LHFP
Gene Name
lipoma HMGIC fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005780.2 2059 UTR 3 NP_005771.1
XM_011534861.1 2059 UTR 3 XP_011533163.1

View Full Product Details