Product Details

SNP ID
rs188270141
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81902513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCGGAGCTGCAACTGCACCCCA[A/G]GCTGCGGAGCCTCGTGAGTCCGGCG
Phenotype
MIM: 614534 MIM: 607996 MIM: 602679 MIM: 606212
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANAPC11 PubMed Links

Gene Details

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

Gene
NPB
Gene Name
neuropeptide B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148896.4 3525 Missense Mutation AAG,AGG K79R NP_683694.1
Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184917.2 3525 Intron NP_001171846.1
NM_001256433.2 3525 Intron NP_001243362.1
NM_001256434.2 3525 Intron NP_001243363.1
NM_001256435.2 3525 Intron NP_001243364.1
NM_001282203.1 3525 Intron NP_001269132.1
NM_001282204.1 3525 Intron NP_001269133.1
NM_002861.4 3525 Intron NP_002852.1
XM_005256386.3 3525 UTR 3 XP_005256443.1
XM_005256387.3 3525 UTR 3 XP_005256444.1
XM_006722287.3 3525 UTR 3 XP_006722350.1
XM_017024910.1 3525 UTR 3 XP_016880399.1
XM_017024911.1 3525 UTR 3 XP_016880400.1
XM_017024912.1 3525 UTR 3 XP_016880401.1
Gene
SIRT7
Gene Name
sirtuin 7
There are no transcripts associated with this gene.

View Full Product Details