Product Details

SNP ID
rs188270862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160508109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGAGCTTATAATAAACTTACCT[C/T]CACATTCTTCTATAAGATTGCCTAT
Phenotype
MIM: 602970 MIM: 615644
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KPNA4 PubMed Links

Gene Details

Gene
KPNA4
Gene Name
karyopherin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002268.4 1676 Missense Mutation GAA,GGA E457G NP_002259.1
Gene
SCARNA7
Gene Name
small Cajal body-specific RNA 7
There are no transcripts associated with this gene.

View Full Product Details