Product Details

SNP ID
rs187952184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140024710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGAGGGGTCGGACACACTGTT[C/T]ACGCAGCTATCATAGAAGGCGTTGC
Phenotype
MIM: 612481 MIM: 274180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP12 PubMed Links

Gene Details

Gene
PARP12
Gene Name
poly(ADP-ribose) polymerase family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022750.3 2830 Silent Mutation GTA,GTG V652V NP_073587.1
XM_005250038.3 2830 Intron XP_005250095.1
XM_005250039.4 2830 Intron XP_005250096.1
XM_005250040.4 2830 Intron XP_005250097.1
XM_011516491.2 2830 UTR 3 XP_011514793.1
Gene
TBXAS1
Gene Name
thromboxane A synthase 1
There are no transcripts associated with this gene.

View Full Product Details