Product Details

SNP ID
rs187860183
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109250266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGGCTCTGTCCATCCTCAGCG[G/T]CGAACCTCTGGCTCTACACCAGCCG
Phenotype
MIM: 604265
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 248 Missense Mutation GCG,TCG A63S NP_001399.1
XM_005270580.4 248 Missense Mutation GCG,TCG A63S XP_005270637.1

View Full Product Details