Product Details

SNP ID
rs188798468
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:43337853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACACAGTGGCGGAGAAGATGC[C/T]CTCCTGGGCCCTCTTCATGGTCACC
Phenotype
MIM: 159530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPL PubMed Links

Gene Details

Gene
MPL
Gene Name
MPL proto-oncogene, thrombopoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005373.2 50 Missense Mutation CCC,CTC P2L NP_005364.1
XM_017001320.1 50 Missense Mutation CCC,CTC P2L XP_016856809.1

View Full Product Details