Product Details

SNP ID
rs188503456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34372398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGCGCCACCATACCAGTGGGC[C/T]GCCGCGTCGCCCAAGAACATGGCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
There are no transcripts associated with this gene.

Gene
KIAA1161
Gene Name
KIAA1161
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020702.4 713 Silent Mutation GCA,GCG A182A NP_065753.2
XM_011517966.2 713 Silent Mutation GCA,GCG A182A XP_011516268.1
XM_017014930.1 713 Silent Mutation GCA,GCG A182A XP_016870419.1

View Full Product Details