Product Details

SNP ID
rs188412281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101726638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAGGGACTACAGAAATTAGGT[A/C]CAAATTTACCCTGTGAAGCTGATAT
Phenotype
MIM: 615864
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEP97 PubMed Links

Gene Details

Gene
CEP97
Gene Name
centrosomal protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303401.1 172 Missense Mutation ACA,CCA T30P NP_001290330.1
NM_024548.3 172 Missense Mutation ACA,CCA T30P NP_078824.2
XM_006713743.3 172 Missense Mutation ACA,CCA T30P XP_006713806.1
XM_011513127.1 172 UTR 5 XP_011511429.1
XM_017007178.1 172 Missense Mutation ACA,CCA T30P XP_016862667.1

View Full Product Details