Product Details

SNP ID
rs188974892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39150360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTCAGTATGTTCTACAGCTTC[C/T]GTATCATCACCTGAAATTTGCCTAA
Phenotype
MIM: 610397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC6B PubMed Links

Gene Details

Gene
TRAPPC6B
Gene Name
trafficking protein particle complex 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079537.1 802 Missense Mutation CAG,CGG Q156R NP_001073005.1
NM_177452.3 802 Missense Mutation CAG,CGG Q128R NP_803235.1
XM_011536409.2 802 Missense Mutation CAG,CGG Q118R XP_011534711.1
XM_017020964.1 802 Intron XP_016876453.1

View Full Product Details