Product Details

SNP ID
rs189172711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136844937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCGGGCCGCTGCGCACCGCCC[A/G]AGCCACCCGCGGGTCCCGCCCCCCA
Phenotype
MIM: 610167 MIM: 610615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf172 PubMed Links

Gene Details

Gene
C9orf172
Gene Name
chromosome 9 open reading frame 172
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080482.2 523 Missense Mutation AAG,GAG K175E NP_001073951.2
Gene
MAMDC4
Gene Name
MAM domain containing 4
There are no transcripts associated with this gene.

Gene
PHPT1
Gene Name
phosphohistidine phosphatase 1
There are no transcripts associated with this gene.

Gene
RABL6
Gene Name
RAB, member RAS oncogene family-like 6
There are no transcripts associated with this gene.

View Full Product Details