Product Details

SNP ID
rs189179469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6891681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGAGTCACTATTGCATAGAAAA[C/T]AGACACCGATTTTTCCTGCTGTTTG
Phenotype
MIM: 608494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 820 Missense Mutation ATT,GTT I274V NP_003691.1

View Full Product Details