Product Details

SNP ID
rs189788782
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26892787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTCTCGGGCATCCTTGTCTCC[A/G]CCATCTGAGGAAACAGAGCTCAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH3 PubMed Links

Gene Details

Gene
GPATCH3
Gene Name
G-patch domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022078.2 1145 Silent Mutation GGC,GGT G372G NP_071361.2
Gene
GPN2
Gene Name
GPN-loop GTPase 2
There are no transcripts associated with this gene.

View Full Product Details