Product Details

SNP ID
rs189926152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10674652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGTCATTCCCTGCAGAGACCA[C/G]AAGGTCGGGAGCGCCTGCGGAGGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C8orf74 PubMed Links

Gene Details

Gene
C8orf74
Gene Name
chromosome 8 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040032.1 185 Missense Mutation CAA,GAA Q19E NP_001035121.1
XM_011543816.2 185 Missense Mutation CAA,GAA Q26E XP_011542118.1
Gene
MIR4286
Gene Name
microRNA 4286
There are no transcripts associated with this gene.

View Full Product Details