Product Details

SNP ID
rs189849720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75478786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCTGCAGCGCACCAGCGCAC[A/C]GTTCCTGCACGCGGCGGATCTTGGC
Phenotype
MIM: 605294
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHST6 PubMed Links

Gene Details

Gene
CHST6
Gene Name
carbohydrate sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021615.4 1838 Missense Mutation CGG,CTG R348L NP_067628.1
XM_005255955.4 1838 Missense Mutation CGG,CTG R348L XP_005256012.1
XM_011523085.2 1838 Missense Mutation CGG,CTG R348L XP_011521387.1

View Full Product Details