Product Details

SNP ID
rs189588511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43028499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGCTCCTCCCGGTCCTCTGAGC[A/G]CAGCGTCAGGGATGTCTTCCATGTG
Phenotype
MIM: 601555 MIM: 604631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RND2 PubMed Links

Gene Details

Gene
RND2
Gene Name
Rho family GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005440.4 450 Missense Mutation CAC,CGC H168R NP_005431.1
XM_011525316.1 450 Missense Mutation CAC,CGC H168R XP_011523618.1
XM_011525317.2 450 Missense Mutation CAC,CGC H140R XP_011523619.1
Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details