Product Details
- SNP ID
-
rs189717919
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21350628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTTTGTGCTGAAGAGGTTGAAG[G/T]TCTGCTGAATCACCTCATGGAGGAC
- Phenotype
-
MIM: 147566
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2988573] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNA6
- Gene Name
- interferon alpha 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021002.2 |
260 |
Missense Mutation |
|
|
NP_066282.1 |
View Full Product Details