Product Details

SNP ID
rs189508421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98383851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTGCTGCATTTCGCCCAGCAG[C/G]TCCCATCACACCTCCTCCTGGAAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PYROXD2 PubMed Links

Gene Details

Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032709.2 1782 Missense Mutation CCT,GCT P565A NP_116098.2
XM_011540293.2 1782 Missense Mutation CCT,GCT P599A XP_011538595.1
XM_011540301.2 1782 Missense Mutation CCT,GCT P296A XP_011538603.1
XM_017016835.1 1782 UTR 3 XP_016872324.1
XM_017016836.1 1782 Intron XP_016872325.1
XM_017016837.1 1782 Intron XP_016872326.1
XM_017016838.1 1782 Missense Mutation CCT,GCT P444A XP_016872327.1
XM_017016839.1 1782 Missense Mutation CCT,GCT P444A XP_016872328.1
XM_017016840.1 1782 Missense Mutation CCT,GCT P444A XP_016872329.1
XM_017016841.1 1782 Missense Mutation CCT,GCT P444A XP_016872330.1

View Full Product Details