Product Details

SNP ID
rs190085149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6892000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAATGCTGTTACTGCCTCGGTA[G/T]GGTAGCCTCAGTATGAAGGTGGTGT
Phenotype
MIM: 608494
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 501 Silent Mutation NP_003691.1

View Full Product Details