Product Details

SNP ID
rs190076880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:658048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCGCTGCGGACCCCGTTTAGTC[C/G]TAACCTCAATCCTGCGAGGGAGGGG
Phenotype
MIM: 616990 MIM: 188350
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLUL1 PubMed Links

Gene Details

Gene
CLUL1
Gene Name
clusterin like 1
There are no transcripts associated with this gene.

Gene
TYMS
Gene Name
thymidylate synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071.2 293 Intron NP_001062.1
Gene
TYMSOS
Gene Name
TYMS opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012716.2 293 Missense Mutation ACG,AGG T67R NP_001012734.2

View Full Product Details