Product Details
- SNP ID
-
rs190228597
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31954637 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGTCTCGATCTCGATCCCGATC[C/T]CGATCCCTGTCCCGCTCTCTGTCTC
- Phenotype
-
MIM: 613927
MIM: 138470
MIM: 154040
MIM: 600478
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs377559803] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2
- Gene Name
- complement component 2
There are no transcripts associated with this gene.
- Gene
- CFB
- Gene Name
- complement factor B
There are no transcripts associated with this gene.
- Gene
- MIR1236
- Gene Name
- microRNA 1236
There are no transcripts associated with this gene.
- Gene
- NELFE
- Gene Name
- negative elongation factor complex member E
- Gene
- SKIV2L
- Gene Name
- Ski2 like RNA helicase
There are no transcripts associated with this gene.
View Full Product Details