Product Details

SNP ID
rs190728401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4989652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTCTTGATCTGATTCAGGGCT[A/C]TTTCCATCAATTTTTCCTGACCAAG
Phenotype
MIM: 605470
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MMP26 PubMed Links

Gene Details

Gene
MMP26
Gene Name
matrix metallopeptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021801.4 125 Missense Mutation TAT,TCT Y35S NP_068573.2
XM_011520219.2 125 UTR 5 XP_011518521.1
Gene
OR51L1
Gene Name
olfactory receptor family 51 subfamily L member 1
There are no transcripts associated with this gene.

View Full Product Details