Product Details

SNP ID
rs190573821
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56346592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAACATAATCAGCTATTTTTACT[A/G]TGACTGTATCCCTCTGATGTCCATA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR8K1 PubMed Links

Gene Details

Gene
OR8K1
Gene Name
olfactory receptor family 8 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002907.1 554 Missense Mutation TAT,TGT Y185C NP_001002907.1

View Full Product Details