Product Details

SNP ID
rs191170812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81645351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACTGGGAGGGCTGGTGGTCAG[C/T]GCAGTGAGCCTGGCTGGCTACCTGG
Phenotype
MIM: 606590 MIM: 180073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPLOC4 PubMed Links

Gene Details

Gene
NPLOC4
Gene Name
NPL4 homolog, ubiquitin recognition factor
There are no transcripts associated with this gene.

Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

Gene
TSPAN10
Gene Name
tetraspanin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290212.1 738 Silent Mutation AGC,AGT S170S NP_001277141.1
NM_031945.4 738 Silent Mutation AGC,AGT S132S NP_114151.3

View Full Product Details