Product Details

SNP ID
rs192086609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44629434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGGAGCAGTTTCTGAGCATC[C/T]TGCCTGGGGAGATTCGGACCTGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF197 PubMed Links

Gene Details

Gene
ZNF197
Gene Name
zinc finger protein 197
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264783.4 469 Silent Mutation CTG,TTG L94L XP_005264840.1
XM_017005495.1 469 Silent Mutation CTG,TTG L94L XP_016860984.1
Gene
ZNF197-AS1
Gene Name
ZNF197 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details